Order Entry
France
ContactUsLinkComponent
 
Toxic
Health
Environment
Arsenic acid
Catalog # 22960.88
CAS Number:  
Toxic
Health
Environment
Arsenic acid
Catalog # 22960.88
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

About this item

 :
 :141.94 g/mol
Merck Index:12,00833
Boiling Pt:160 °C (1013 hPa)
Melting Pt:35.5 °C
Density:2.2 g/cm³ (20 °C)
EINECS:231-901-9
UN:1553
ADR:6.1,I
 :GHS06
 :GHS08
 :GHS09
CAS Number:7778-39-4
Formula:H₃AsO₄