Order Entry
France
ContactUsLinkComponent
Sodium hydroxide 1% in aqueous solution
Corrosive
Sodium hydroxide 1% in aqueous solution
Catalog # PRLSMC310001.9025
Supplier:  PROLABO CMP
CAS Number:  
Corrosive
Sodium hydroxide 1% in aqueous solution
Catalog # PRLSMC310001.9025
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 L
  • Seq:
    5327

Specifications

About this item

 :
 :40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548