
Benzyl chloroacetate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140-18-1
:
Formula:C₉H₉ClO₂
MDL Number:MFCD00157062
:184.62 g/mol
Boiling Pt:132 °C (12 mmHg)
Density:1.22 g/cm³
Flash Pt:>110 °C
EINECS:205-400-0
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5240 to 1.5270 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together