3-Cyano-5-methylpyridine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:42885-14-3
:
Formula:C₇H₆N₂
MDL Number:MFCD00673151
:118.14 g/mol
Boiling Pt:220 °C (1013 hPa)
Melting Pt:82…85 °C
Storage Temperature:Ambient
EINECS:255-988-8
:GHS07
Related Information
Frequently Bought Together