Mucic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:526-99-8
:
Formula:C₆H₁₀O₈
MDL Number:MFCD00004239
Melting Pt:230 °C (dec.)
Density:1,79 g/cm³ (20 °C)
:210.14 g/mol
EINECS:208-404-0
Specification Test Results
- Appearance:White to light beige Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 %
- Sulfated ash:≤0.2 %
- Titration excess NaOH:≥97.5 %
- Barium (Ba):≤0.005 %
- Calcium (Ca):≤0.001 %
- Lithium (Li):≤0.001 %
- Magnesium (Mg):≤0.0002 %
- Potassium (K):≤0.03 %
- Rubidium (Rb):≤0.005 %
- Sodium (Na):≤0.02 %
- Strontium (Sr):≤0.0005 %
Specification Test Results
Frequently Bought Together