Hydrocortisone, Millipore®
Catalog # 386698-25
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:362.47 g/mol
Boiling Pt:567 °C (1013 hPa)
Melting Pt:202…204 °C
Density:1.4 g/cm³ (20 °C)
EINECS:200-020-1
CAS Number:50-23-7
Formula:C₂₁H₃₀O₅
MDL Number:MFCD00011654