
1,1,1-Trifluoro-3-phenylacetone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:350-92-5
:
Formula:C₉H₇F₃O
MDL Number:MFCD00068171
:188.15 g/mol
Boiling Pt:51…52 °C (2 mmHg)
Density:1.220
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together