Order Entry
France
ContactUsLinkComponent
Accessories for aspirating pipettes, Costar®
  734-1998
 :  Corning
undefined
Accessories for aspirating pipettes, Costar®
  734-1998
 :  Corning
  • Description:
    8-channel adapter for use with Costar® vacuum aspirator
  • Pk:
    1
This is an 8-channel adapter that fits on the Costar® aspirator so that it can be used to remove media and solutions from 8 wells simultaneously on a 96 well plate.