Order Entry
France
ContactUsLinkComponent
Magnesium chloride-4,5-hydrate ≥97%, powder, GPR RECTAPUR®
Magnesium chloride-4,5-hydrate ≥97%, powder, GPR RECTAPUR®
Catalog # 25106.298
Supplier:  VWR Chemicals
CAS Number:  
undefined
Magnesium chloride-4,5-hydrate ≥97%, powder, GPR RECTAPUR®
Catalog # 25106.298
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 97 %
  • Alkaline metals (as SO4):
    Max. 2.5 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Ca (Calcium):
    Max. 0.1 %
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :176.28 g/mol
Boiling Pt:1412 °C (1013 hPa)
Density:1.57 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-094-6
 :7791-18-6
Formula:MgCl₂·4,5H₂O