Order Entry
France
ContactUsLinkComponent
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Toxic
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Barium chloride dihydrate ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • pH (25°C; 5 %):
    5.2 - 8.2
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Loss on drying (150°C):
    14.0 - 16.0 %
  • Total N (Nitrogen):
    Max. 20 ppm
  • Ca (Calcium):
    Max. 50 ppm
  • Fe (Iron):
    Max. 1 ppm
  • K (Potassium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Pb (Lead):
    Max. 10 ppm
  • Sr (Strontium):
    Max. 0.05 %
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

Frequently Bought Together