Order Entry
France
ContactUsLinkComponent
Disodium hydrogen phosphate heptahydrate 98+% ACS
Disodium hydrogen phosphate heptahydrate 98+% ACS
Certificates
undefined
Disodium hydrogen phosphate heptahydrate 98+% ACS

Specification Test Results

  • Appearance:
    Colorless to white Granular powder or crystals
  • pH:
    8.7 to 9.3 (5 % solution at 25.0°C)
  • Titration excess HCl:
    98.0 to 102.0 %
  • Chloride (Cl):
    ≤10 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

Related Information

Frequently Bought Together