Order Entry
France
ContactUsLinkComponent
 
Citric acid monohydrate ≥99.9%, tested according to Ph. Eur., Fluka™
 
undefined
Citric acid monohydrate ≥99.9%, tested according to Ph. Eur., Fluka™

 

  • Identity (IR):
    complying
  • Assay (calculated on the anhydrous basis):
    99.5 - 100.5 %
  • Appearance of the solution:
    complying
  • Sulfated ash:
    Max. 0.1 %
  • Water (Karl Fischer):
    7.5 - 9.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 150 ppm
  • Reaction against H₂SO₄:
    complying
  • Readily carbonisable substances:
    complying
  • Oxalate (as Oxalic acid):
    Max. 360 ppm
  • Residual solvents:
    complying

 

 

Frequently Bought Together