Order Entry
France
ContactUsLinkComponent
Silver nitrate 0.025 mol/l (0.025 N) in aqueous solution
Silver nitrate 0.025 mol/l (0.025 N) in aqueous solution
Catalog # PRLSMC306825.5000
Supplier:  PROLABO CMP
CAS Number:  
undefined
Silver nitrate 0.025 mol/l (0.025 N) in aqueous solution
Catalog # PRLSMC306825.5000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414