Sodium thiosulphate 1 mol/l (1 N) in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:7772-98-7
:
Formula:Na₂S₂O₃
MDL Number:MFCD00003499
:158.11 g/mol
Storage Temperature:Ambient
EINECS:231-867-5
Frequently Bought Together