
Sodium hydroxide 2 mol/l (2 N) in aqueous solution
Supplier: PROLABO CMP
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:≤1.11 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:GHS05
Frequently Bought Together