Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
France
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium hydroxide
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
PRLSMC310208.9050
:
PROLABO CMP
:
Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
PRLSMC310208.9050
:
PROLABO CMP
:
Pack type:
Plastic drum
Pk:
50 L
Seq:
5377
:
:
40 g/mol
:
1 g/cm³ (25 °C)
:
Ambient
:
215-185-5
:
1824
:
8,III
:
1310-73-2
:
NaOH
:
MFCD00003548