Order Entry
France
Orders LinkContactUsLinkComponent
 

(±)-2-Hexanol ≥99%

Flammable
Irritant
(±)-2-Hexanol ≥99%
Certificates
(±)-2-Hexanol ≥99%
(±)-2-Hexanol ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 626-93-7 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 45°(113°F) Refractive Index: 1.4140 Odour: Characteristic Melting Point: -23° Boiling Point: 137-139° Beilstein Reference: 1718996 UN No.: 2282 MDL No.: MFCD00004585

Synonyms:  (±)-Methyl butyl carbinol, (±)-Butyl methyl carbinol, (±)-2-Hydroxy hexane
  • CAS Number:626-93-7
  • Formula:C₆H₁₄O
  • MDL Number:MFCD00004585
  • Molecular Weight:102.18 g/mol
  • Boiling Pt:137…139 °C
  • Melting Pt:–23 °C
  • Density:0.810
  • Flash Pt:45 °C (113 °F)
  • Storage Temperature:Ambient
  • EINECS:210-971-4
  • UN:2282
  • ADR:3,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.