Order Entry
France
ContactUsLinkComponent
Perchloric acid 60%, AnalaR NORMAPUR® analytical reagent
Perchloric acid 60%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Perchloric acid 60%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    60 - 62 %
  • Free chlorine:
    Max. 5 ppm
  • Ignition residue (SO4):
    Max. 30 ppm
  • Total N (Nitrogen):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 3 ppm
  • ClO3 (Chlorate):
    Max. 10 ppm
  • PO4 + SiO4 (as SiO2):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 10 ppm
  • Ag (Silver):
    Max. 5 ppm
  • As (Arsenic):
    Max. 0.05 ppm
  • Cu (Copper):
    Max. 0.1 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Mg (Magnesium):
    Max. 10 ppm
  • Mn (Manganese):
    Max. 0.5 ppm
  • Pb (Lead):
    Max. 0.1 ppm

Specification Test Results

Frequently Bought Together