


Formaldehyde 36% in aqueous solution Ph. Eur.
Supplier: VWR Collection
Certificates
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
Storage Temperature:Ambient
EINECS:200-001-8
UN:2209
ADR:8,III
:GHS06
:GHS05
:GHS08
Related Information
Certificates
Search by Lot Numberspec_VWRS30322.368 spec_VWRS30322.368 spec_VWRS30322.465 spec_VWRS30322.465
Product specification
Frequently Bought Together