Order Entry
France
ContactUsLinkComponent
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Certificates
undefined
Dichloromethane-[D₂] 99.8 atom % D for NMR spectroscopy
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.02 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %