Sodium chloride : Sodium hydroxide (175:150) g/l in aqueous solution
Catalog # VWRS83959.460
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:25 L
- Seq:5327
- Environmentally Preferable:
Specifications
About this item
:
Density:1.19 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3266
ADR:8,II
:GHS05