Order Entry
France
ContactUsLinkComponent
May Grunwald's eosin-methylene blue
Irritant
May Grunwald's eosin-methylene blue
Catalog # FFCHFF0911000
Supplier:  FF CHEMICALS
CAS Number:  
Irritant
May Grunwald's eosin-methylene blue
Catalog # FFCHFF0911000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00131580