
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:194491-32-2
:
Formula:C₁₀H₁₂MgN₂Na₂O₈·xH₂O
MDL Number:MFCD00078217
:358.50 g/mol (anhy)
Melting Pt:>300 °C
EINECS:238-372-3
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together