VWR® Software for Validation of Analytical Methods, Validation Manager
HPLC1.18988.SMA1
- Description:SMA (Software Maintenance Agreement) Validation Manager, single-user license
- Environmentally Preferable:
- Pk:1
Validation manager software checks whether the selected analytical method is suitable for the use intended, and automatically produces the required validation report. As a result, validation manager can save days or even weeks of validation work for any analytical technique.
- Based on the international guidelines for method validation
- Versatile configuration for all analytical techniques
- Ease of operation and easy data entry
- Automatic calculation and evaluation of the method characteristics
- Automatic report compilation
- Validated and fully FDA compliant
For further information on Validation Manager software, please contact the specialists at Avantor or email [email protected].