Order Entry
France
Orders LinkContactUsLinkComponent
HPLC standards, LiChroTest®, Supelco®
HPLC standards, LiChroTest®, Supelco®
HPLC standards, LiChroTest®, Supelco®
Supplier:  Merck
Certificates
6 Options Available Below
Product Details & Documents
Standard samples for HPLC System Qualification.
In order to facilitate this instrument qualification in the HPLC laboratory Merck Millipore and Avantor have developed the LiChroTest® products. These enable time saving Operational and Performance Qualification to be performed routinely and according to standardised methods.
The quality of analytical results is principally determined by the correct functioning of the analytical instruments employed. For this reason, the various quality assurance systems as well as the FDA require analytical instruments to be subjected to periodic qualification. Therefore, before starting a series of analyses, you first should establish whether your HPLC system meets with your requirements. These 'Operational Qualification' (OQ) and 'Performance Qualification' (PQ) steps involve tests of the different modules on their specifications and a check on the entire system using a real application relevant to the laboratory-specific requirements.
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 909-0002
  • 909-0004
  • 909-0005
  • 909-0007
  • 909-0008
  • 909-0009
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
Specifications
Product Family Options
LiChroTest® standard samples for Performance Qualification (PQ) of HPLC systems
Product Information
  • Pk
    Availability
    Price
  • 909-0002
    Set 1A: Precision and Linearity (PQ), refill pack for the LiChroTest® PQ Kit. 1.19156.0001, dilution series of methyl paraben in methanol/water (50/50) (conc.: 50, 100, 150, 200 mg/l)
    Set 1A: Precision and Linearity (PQ), refill pack for the LiChroTest® PQ Kit. 1.19156.0001, dilution series of methyl paraben in methanol/water (50/50) (conc.: 50, 100, 150, 200 mg/l)
    5
    Log in for availability
    Log in for availability
    1 * 5 Pce
    1 * 5 Pce - 419,00 €
    419,00 €
    1 * 5 Pce
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    909-0002
    M-Clarity quality level
    MQ100
  • 909-0004
    Set 2: Precision (PQ), 5 Standard samples of 100 mg/l methyl paraben in methanol/water (50/50), refill pack for the LiChroTest® PQ set
    Set 2: Precision (PQ), 5 Standard samples of 100 mg/l methyl paraben in methanol/water (50/50), refill pack for the LiChroTest® PQ set
    5
    Log in for availability
    Log in for availability
    1 * 5 Pce
    1 * 5 Pce - 419,00 €
    419,00 €
    1 * 5 Pce
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    909-0004
    M-Clarity quality level
    MQ100
  • 909-0005
    Set 3: Separation (Parabens) (PQ), 5 standard samples with 3 different parabens + t₀-marker in methanol/water (50/50), with sample chromatogram and analysis conditions
    Set 3: Separation (Parabens) (PQ), 5 standard samples with 3 different parabens + t₀-marker in methanol/water (50/50), with sample chromatogram and analysis conditions
    5
    Log in for availability
    Log in for availability
    1 * 5 Pce
    1 * 5 Pce - 419,00 €
    419,00 €
    1 * 5 Pce
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    909-0005
    M-Clarity quality level
    MQ100
LiChroTest® standard samples for Operational Qualification (OQ) of HPLC systems
Product Information
  • Pk
    Availability
    Price
  • 909-0007
    Set 7: Precision (OQ), 60 mg/l methyl paraben in methanol
    Set 7: Precision (OQ), 60 mg/l methyl paraben in methanol
    1
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 419,00 €
    419,00 €
    1 * 1 Pce
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    909-0007
    M-Clarity quality level
    MQ100
  • 909-0008
    Set 8: Linearity (OQ), dilution series of methyl paraben in methanol (conc.: 1,5; 7,5; 15; 75; 150 mg/l))
    Set 8: Linearity (OQ), dilution series of methyl paraben in methanol (conc.: 1,5; 7,5; 15; 75; 150 mg/l))
    1
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 419,00 €
    419,00 €
    1 * 1 Pce
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    909-0008
    M-Clarity quality level
    MQ100
  • 909-0009
    Caffeine solution A for gradient test (OQ), 20 mg/l caffeine in water (0,5 l)
    Caffeine solution A for gradient test (OQ), 20 mg/l caffeine in water (0,5 l)
    1
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 419,00 €
    419,00 €
    1 * 1 Pce
       419,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    909-0009
    M-Clarity quality level
    MQ100