Taps
About this item
The taps are of 3 types:
- Taps for hard and demineralised water
- Taps for combustible gases
- Taps for air, compressed gas and vacuum
Straight hose length 80 cm, exterior stainless steel braid, interior neoprene sheath. Suitable for taps on the table-top (for cold water, hot water, compressed air). Outlet by 15/21 female nut for connection on 15/21 (¹/₂") male standby gas feed.
Elbow hose with same characteristics as straight hoses but adapted to plumbing fittings on splash back (for cold water, hot water, compressed air).
Ordering information: Different sockets (individual and in a row) are available on request.
Caution: * All of these valves are available in a counter-top version.
11 Options Available Below
Product Details & Documents
The taps are of 3 types:
- Taps for hard and demineralised water
- Taps for combustible gases
- Taps for air, compressed gas and vacuum
Straight hose length 80 cm, exterior stainless steel braid, interior neoprene sheath. Suitable for taps on the table-top (for cold water, hot water, compressed air). Outlet by 15/21 female nut for connection on 15/21 (¹/₂") male standby gas feed.
Elbow hose with same characteristics as straight hoses but adapted to plumbing fittings on splash back (for cold water, hot water, compressed air).
Ordering information: Different sockets (individual and in a row) are available on request.
Caution: * All of these valves are available in a counter-top version.
Product Family Options
Product Information
- AvailabilityPrice
- 132-2705Flexible PVC evacuation for waste water, diam. 40mm
- 132-2706Straight supply hose, length 80 mm, On table
- 132-2707Hot/cold water mixer tap, for tank, On backsplash*
- 132-2711Adjustable gooseneck cold water tap, for tank, On backsplash*Adjustable gooseneck cold water tap, for tank, On backsplash*Log in for availability1 * 1 Pce176,00 €
- 132-2712Direct-operated countertop faucet for sink, Countertop
- 132-2709Combustible gas tap, On backsplash
- 132-2714Robinet gaz combustible, Sur table
- 132-2713Compressed air tap, On backsplash
- 132-2708Compressed Air Tap, Tabletop
- 132-2710Demineralised water gooseneck tap, for font, On backsplash*Demineralised water gooseneck tap, for font, On backsplash*Log in for availability1 * 1 Pce201,00 €
- 132-2704Elbow hose, On table