Order Entry
France
Orders LinkContactUsLinkComponent
Overhead cabinets

Overhead cabinets

Sliding doors, glazed on 2 sides *, 900×350×685 mm
Catalog # 132-2723
Supplier:   EQUIP LABO
Overhead cabinets
Catalog # 132-2723
Supplier:  EQUIP LABO
Overhead cabinets
Overhead cabinets

Sliding doors, glazed on 2 sides *, 900×350×685 mm

Price
1 230,00 €
Log in for availability
1 * 1 Pce
Product Details & Documents

Supplied assembled with fitting accessories, length 900 mm.

Specifications
  • Description:
    Sliding doors, glazed on 2 sides *
  • W×D×H:
    900×350×685 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.