Order Entry
France
ContactUsLinkComponent
 
behrotest® DET supplementary set for Soxhlet extraction (100 ml) DE 100
  701-0069
 :  BEHR
behrotest® DET supplementary set for Soxhlet extraction (100 ml) DE 100
  701-0069
 :  BEHR

 

  • Type:
    DE 100
  • Description:
    Soxhlet extractor 100 ml set
  • Pk:
    1

 

 

Soxhlet extraction designed for larger solid samples with five sample positions.

This set includes the following equipment:
5×Sample vessels, 250 ml 65/250
5×Vessel –extractor connector M 2965
5×100 ml soxhlet extractor EZ 100
5×Reflux condenser for EZ 100 extractor RFK 100

 : The DET Basic unit (DBAS, 705-0023) is also required.