Dispensers for cleanroom wipes
Supplier: S-CURVE
Certificates
About this item
Made of clear 6 mm thick acrylic, with hinged lid.
- Protects wipes against contamination
- Full-length hinged lid keep wipes clean and easily accessible for filling
- Flame treated edges for smoothness
- Wall mountable or free-standing with rubber pads on bottom corners
Frequently Bought Together