Flange kits, Easy-Flange
Catalog # 554-1042
Supplier: VICI JOUR RESEARCH
Specifications
- Description:Easy flange kits
- Pk:1
Specifications
About this item
The Easy-Flange flange-rolling tool has been developed for flanging all PTFE and PTFE-like tubes with Ext.Ø between ¹/₁₆' and ¹/₈' where fittings requiring a flange.
- Handy tool for perfect flanges
- Tool forms the flange by mechanical force, no heat or electricity required
- No contamination
Delivery: The kit contains flanging disc with 0,5 mm SS pin for PEEK tubing; flanging disc with 0,8 mm polymer pin; flanging disc with 0,8 mm titanium pin; flanging disc with 1,3 mm polymer pin; flanging disc with 1,3 mm titanium pin; clean-cut tubing cutter; PTFE tubing ¹/₁₆'×0,75 mm×3 m.