Chromatography tubing
SS316 welded under inert gas.
- Smooth internal surface
- Soft annealed OD for easy ferrule connections
A variety of sizes for HPLC and GC applications. Tubings can withstand a pressure rating of min 315 bar (4600 psi.).
Frequently Bought Together