Culture slides, Falcon®
Supplier: Corning
Certificates
About this item
Soda-lime glass slide, PS vessel, lid and tool for loosening chamber.
- Chamber easily and safely removed using the disposable safety removal tool
- Pressure-sensitive, biocompatible, acrylic-adhesive gasket remains with the vessel after removal, not on the slide, facilitating further processing or placement of coverslips
- Blue hydrophobic border defines cell culture areas
- Wells numbered for easy identification
- Trays designed for incubator use
Falcon® CultureSlides allow cells to be cultured and then analysed on a glass microscope slide. Cells are grown in a plastic chamber attached to a specially prepared microscope slide. Cells can be fixed and stained in situ without disruption of the cell monolayer.
W×L: 25×75 mm with 1,2 mm bevelled edge.
Ordering information: Supplied with removal tool and key.
Frequently Bought Together