Well caps for U96 DeepWell™ plates, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Made from thermoplastic elastomer (TPE), except pre-slit silicone well cap. For sealing 96-well plates, particularly 1,3 ml and 2,0 ml U96 DeepWell™ plates. TPE well caps are resistant to DMSO, ethanol and methanol. Silicone well cap is solvent resistant, but is not suitable for 100% DMSO.
- Suitable for plates being used for chemical and compound storage
- Produces an efficient seal with minimum protrusion into each well
- Pierceable and self-closing
- Can be used at temperatures down to −80 °C (except silicone cap, −20 °C)
Specifications
- Catalog No:
- 732-2668
- 732-2669
- 732-2672
- 732-2673
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- NAR Certificate of Retirement