Plates, 1536-well, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
About this item
PS, non treated, without lid. Designed for ultra-high throughput screening applications.
- Standard microplate footprint
- Available non treated in black, white or clear
- Low profile lid available separately
Working range: 1 to 10 µl.
Specifications
- Catalog No:
- 732-2710
- 732-2711
- 732-2709
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement