MicroWell™ Plates, 96 well, Nunc™
Supplier: Thermo Fisher Scientific
Certificates
About this item
PS, clear, untreated. Used in many fields including antibiotic screening, serological testing, storing and screening of compounds, and DNA libraries.
- Raised well rims and corresponding condensation rings on the lid prevent cross-contamination and evaporation
- Optimised for robotics, automated readers and liquid handling systems
- Flat bottom shape is ideal for microscopic and optical measurements
- Conical well shape maximises sample recovery
- Barcoded version also available
Specifications
- Catalog No:
- 734-2611
- 732-2717
- 732-2720
- 732-2747
- 732-2721
- 732-2718
- 732-2719
- 732-2744
- 732-2715
- 732-2725
- 738-0147
- 732-0191
- 734-0491
- 732-0812
- 732-2746
- 738-1003
- 738-1004
- 732-2702
- 738-0196
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Certificate Registration No.CERT-IT000330)
- NAR Certificate of Retirement