Commercial grade hydrometers
Supplier: Alla France
Certificates
About this item
Commercial grade hydrometers − Not standardised.
- Reference mark on the stem
- Check of the calibration on the paper scale
- Bulb Ø is 15 or 20 mm with a stem Ø of 6 to 7 mm for greater accuracy
Frequently Bought Together