Order Entry
France
ContactUsLinkComponent
Intensive regenerating hand creams, Baktolan® Balm Intensive Care
Intensive regenerating hand creams, Baktolan® Balm Intensive Care
  BODE972553
undefined
Intensive regenerating hand creams, Baktolan® Balm Intensive Care
  BODE972553
  • Description:
    Intensive regenerating hand cream, Baktolan® Balm Intensive Care
  • Pk:
    350 mL
Baktolan® balm is a rich hand care cream for daily use on dry and sensitive skin. Due to its replenishing characteristics, Baktolan® balm is used wherever sensitive or vulnerable skin needs care.
  • W/O emulsion for the care of dry skin and sensitive skin
  • Contains allantoin, vitamin E, panthenol, and bisabolol
  • Alleviates skin irritations and redness
  • Stabilises the protective acid mantle
  • Absorbes without leaving a greasy film
Suitable for daily use in utility departments and cleaning in all areas of health care, industry, and laboratories.
 : * These articles are only available in Germany.
*The product(s) on this page are outside the scope of the EU Directive 2019/904