Order Entry
France
Orders LinkContactUsLinkComponent
3-piece filter funnels, Whatman™
3-piece filter funnels, Whatman™
3-piece filter funnels, Whatman™
Certificates
3-piece filter funnels, Whatman™
3-piece filter funnels, Whatman™
7 Options Available Below
Product Details & Documents
Whatman 3-piece filter funnels from Cytiva are ideal for quick and easy filtration using high-efficiency glass microfiber filters.
  • Chemically resistant - 3-piece construction consists of glass cylinder and base with a polymeric membrane support
  • Support options - Provided with acrylic support and optional polypropylene or PTFE supports available separately for some sizes
  • Secure sealing - Clamps at edges to prevent peripheral loss due to fluid bypass around the edge of the filter
  • Eliminate carry-over - Simplicity of design facilitates easy and thorough cleaning between use
The increased use of high-efficiency glass microfiber filters in modern laboratories has created a demand for simple and effective filter-holding systems. Whatman 3-piece Filter Funnels have been designed to complement the range of Whatman fine particle retention, rapid flow rate glass microfiber filters.
The unit consists of a glass funnel and base, acrylic support platform, and 3 easy-to-apply metal clips. The range of diameters, funnel sizes, reservoir volumes and alternate support platforms provides a flexible system to meet the varied and unique needs of busy laboratories.
Product Family Options
Product Information
  • Capacity
    Plate Ø
    For filter Ø
    Availability
    Price
  • 16 ml
    30 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 240,00 €
    240,00 €
    1 * 1 Pce
       240,00 €
  • 36 ml
    47 mm
    47 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 272,00 €
    272,00 €
    1 * 1 Pce
       272,00 €
  • 115 ml
    70 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 331,00 €
    331,00 €
    1 * 1 Pce
       331,00 €
  • 210 ml
    70 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 582,00 €
    582,00 €
    1 * 1 Pce
       582,00 €
  • 400 ml
    70 mm
    70 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 572,00 €
    572,00 €
    1 * 1 Pce
       572,00 €
  • 200 ml
    90 mm
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 583,00 €
    583,00 €
    1 * 1 Pce
       583,00 €
  • 530 ml
    125 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 697,00 €
    697,00 €
    1 * 1 Pce
       697,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Filter funnel plate, PP, 90 mm
    Filter funnel plate, PP, 90 mm
    511-1000
    Log in for availability
    1 * 1 Pce - 107,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter funnel plate, PTFE, 47 mm
    Filter funnel plate, PTFE, 47 mm
    WHAT1950-114
    Log in for availability
    1 * 1 Pce - 102,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter funnel plate, PTFE, 70 mm
    Filter funnel plate, PTFE, 70 mm
    WHAT1950-117
    Log in for availability
    1 * 1 Pce - 130,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter funnel replacement reservoir, 115 ml, 70 mm
    Filter funnel replacement reservoir, 115 ml, 70 mm
    WHAT1950-207
    Log in for availability
    1 * 1 Pce - 156,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter funnel replacement reservoir, 200 ml, 90 mm
    Filter funnel replacement reservoir, 200 ml, 90 mm
    WHAT1950-209
    Log in for availability
    1 * 1 Pce - 181,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filter funnel replacement reservoir, 210 ml, 70 mm
    Filter funnel replacement reservoir, 210 ml, 70 mm
    516-0612
    Log in for availability
    1 * 1 Pce - 197,00 €