Tubing connectors, mini, Y-shape
229-0501
- Tubing int.Ø:1,6 mm
- Pk:10
PP, translucent.
- Very good thermal resistance
- Very good chemical resistance
- Robust, durable
Perfect for conneting tubing with small inner diameters
PP, translucent.
Perfect for conneting tubing with small inner diameters