Jars, wide mouth, with screw cap
Supplier: KARTELL
Certificates
About this item
HDPE, translucent.
- Good chemical resistance
- Temperature resistant from –50 to +80 °C
- Wide opening makes it ideally suited for storing and transporting powders and pastes
With black screw cap and sealing element.
Certifications: Suitable for foodstuff according to EC regulation 10/2011.
Limited Time Sale
Select items below are currently on sale