Carboys, wide neck, PP or PC, with screw closure and spigot, Nalgene®
PP, translucent with white PP screw cap and PP spigot with TPE seal, or PC, clear with white PP screw cap and PP spigot with TPE seal.
- PP: Very good chemical resistance, ideal for storing, transporting and dispensing solutions, culture media and distilled water/PC: Good chemical resistance, ideal for storing, transporting and dispensing sterile water, standard reagents and culture media
- PP: Temperature resistant from –10 to +135 °C, autoclavable/PC: Temperature resistant from –100 to +135 °C, autoclavable
- Leakproof closure
Wide mouth opening for easy filling and cleaning. For better durability and leak resistance the drain cock thread is moulded in one piece with the carboy, not welded on. Graduated.
- Catalog No:
- 216-8792
- 216-8791
- 216-4411
- 216-8772
- 216-8771
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together