Order Entry
France
Orders LinkContactUsLinkComponent
Aluminium foil discs
Supplier:   Korff
Aluminium foil discs
Supplier:  Korff
Certificates
Aluminium foil discs
Aluminium foil discs
15 Options Available Below
Product Details & Documents
Pure aluminium, 99,0 to 99,5%.
  • Economical large packs
  • Tissue paper is inserted between discs to facilitate handling
  • Simple storage and removal from dispenser
Can be used for a wide variety of laboratory tasks.
Product Family Options
Product Information
  • Thickness
    Ø
    Availability
    Price
  • 10 µm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 111,00 €
    111,00 €
    1 * 1.000 Pce
       111,00 €
  • 10 µm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 154,00 €
    154,00 €
    1 * 1.000 Pce
       154,00 €
  • 20 μm
    50 mm
    Log in for availability
    Log in for availability
    1 * 500 Pce
    1 * 500 Pce - 128,00 €
    128,00 €
    1 * 500 Pce
       128,00 €
  • 30 µm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 455,00 €
    455,00 €
    1 * 1.000 Pce
       455,00 €
  • 30 µm
    220 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 484,00 €
    484,00 €
    1 * 1.000 Pce
       484,00 €
  • 30 µm
    29 mm
    Log in for availability
    Log in for availability
    1 * 5.000 Pce
    1 * 5.000 Pce - 784,00 €
    784,00 €
    1 * 5.000 Pce
       784,00 €
  • 30 µm
    80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 95,00 €
    95,00 €
    1 * 1.000 Pce
       95,00 €
  • 30 µm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 120,00 €
    120,00 €
    1 * 1.000 Pce
       120,00 €
  • 30 µm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 162,00 €
    162,00 €
    1 * 1.000 Pce
       162,00 €
  • 30 µm
    130 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 206,00 €
    206,00 €
    1 * 1.000 Pce
       206,00 €
  • 30 µm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 234,00 €
    234,00 €
    1 * 1.000 Pce
       234,00 €
  • 30 µm
    110 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 273,00 €
    273,00 €
    1 * 1.000 Pce
       273,00 €
  • 30 µm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 390,00 €
    390,00 €
    1 * 1.000 Pce
       390,00 €
  • 50 µm
    225 mm
    Log in for availability
    Log in for availability
    1 * 1.000 Pce
    1 * 1.000 Pce - 573,00 €
    573,00 €
    1 * 1.000 Pce
       573,00 €
  • 50 µm
    100 mm
    Log in for availability
    Log in for availability
    1 * 500 Pce
    1 * 500 Pce - 137,00 €
    137,00 €
    1 * 500 Pce
       137,00 €