Order Entry
France
ContactUsLinkComponent
Mercury standard solution, 1000 mg/L Hg in 10% nitric acid standard for ICP
Mercury standard solution, 1000 mg/L Hg in 10% nitric acid standard for ICP
Catalog # JOBIJYICP-HG
CAS Number:  
Mercury standard solution, 1000 mg/L Hg in 10% nitric acid standard for ICP
Catalog # JOBIJYICP-HG
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item