EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-42-5
:
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291
Merck Index:14,3529
Melting Pt:∼ 242 °C (dec.)
:380.35 g/mol
Storage Temperature:Ambient
EINECS:200-651-2
Related Information
Frequently Bought Together