Order Entry
France
ContactUsLinkComponent
Human Recombinant IL-34 (from CHO cells)
Human Recombinant IL-34 (from CHO cells)
New Product
Certificates
Human Recombinant IL-34 (from CHO cells)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0,1
  • Biological activity:
    Bioactivity assay: Currently undetermined, ED50 ≤Currently undetermined, Bioactivity ≥NA
  • Gene ID:
    NP_689669
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml (reconstitution at concentrations > 0.1 mg/ml may result in a precipitate)
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein/peptide name:
    IL-34
  • Purity:
    ≥95%
  • Molecular weight:
    26 kDa
  • Sequence:
    NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP KVESVLSLLN APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHH
  • Endotoxin level:
    Low
  • Formulation:
    Phosphate Buffered Saline (PBS)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together