Order Entry
France
ContactUsLinkComponent
Ninhydrin and stannous chloride reagent Reag. Ph. Eur. 1058301
Ninhydrin and stannous chloride reagent Reag. Ph. Eur. 1058301
Catalog # 87875.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Ninhydrin and stannous chloride reagent Reag. Ph. Eur. 1058301
Catalog # 87875.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Storage Temperature:Ambient