0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
France
ContactUsLinkComponent
Potassium permanganate
Potassium permanganate 0.2 mol/l (1 N) in aqueous solution
Potassium permanganate 0.2 mol/l (1 N) in aqueous solution
FFCHFF0801000
:
FF CHEMICALS
:
Potassium permanganate 0.2 mol/l (1 N) in aqueous solution
FFCHFF0801000
:
FF CHEMICALS
:
Pack type:
Plastic bottle
Pk:
1 L
Seq:
5037
:
:
158.03 g/mol
:
Ambient
:
231-760-3
:
3082
:
9,III
:
GHS09
:
7722-64-7
:
KMnO₄
:
MFCD00011364