Order Entry
France
ContactUsLinkComponent
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution
Environment
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF0760500
Supplier:  FF CHEMICALS
CAS Number:  
Environment
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF0760500
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :158.03 g/mol
Storage Temperature:Ambient
EINECS:231-760-3
UN:3082
ADR:9,III
 :GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364