VWR®, Conductivity standard 100000 µS/cm (25 °C; 0,85 mol/L; KCl) for conductivity measurement
Catalog # VWRS38492.0500
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Reference temperature:25 °C
- Description:Conductivity standard 100,000 µS/cm (0,85 mol/l; KCl)