Order Entry
France
ContactUsLinkComponent
Primary solution yellow (1 ml solution contains 45.0 mg FeCl3.6H2O) Reag. Ph. Eur.
Primary solution yellow (1 ml solution contains 45.0 mg FeCl3.6H2O) Reag. Ph. Eur.
  PRLS87985.180
 :  
Primary solution yellow (1 ml solution contains 45.0 mg FeCl3.6H2O) Reag. Ph. Eur.
  PRLS87985.180
 :  
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    Primary solution yellow (1 ml solution contains 45.0 mg FeCl3.6H2O) Reag. Ph. Eur.